missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. p-5383259
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02002848 500 mL Narrow Mouth Amber Glass Bottle
02002847 4 L Narrow Mouth PVC-coated Amber Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02002848 Supplier Avantor Supplier No. 930001
Only null left

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Quantity 500 mL
Flash Point 111.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
Formula Weight 62.07
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Ethylene Glycol