Learn More
Ethylene glycol, 99.5%, for analysis
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Supplier: Thermo Scientific Chemicals 295530025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
| Ethylene glycol | |
| 107-21-1 | |
| 196.0°C to 198.0°C | |
| 116°C | |
| Glass Bottle | |
| 1.4300 to 1.4330 | |
| 1.113 | |
| HOCH2CH2OH | |
| 21 mPa.s (20°C) | |
| 01, 465 | |
| 15, 3852 | |
| Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons | |
| C(CO)O | |
| 62.068 | |
| CHEBI:30742 | |
| 99.5% |
| -13.0°C | |
| 1.1130g/mL | |
| 2.5 L | |
| Authentic | |
| Viscous Liquid | |
| 99.5% | |
| C2H6O2 | |
| MFCD00002885 | |
| 0.1% max. (K.F.) | |
| 01,375; 05,296; 09,217; 15,156 | |
| ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| ethane-1,2-diol | |
| 174 | |
| 62.06 | |
| Analytical |
Chemical Identifiers
| 107-21-1 | |
| 62.068 | |
| LYCAIKOWRPUZTN-UHFFFAOYSA-N | |
| 174 | |
| ethane-1,2-diol |
| C2H6O2 | |
| MFCD00002885 | |
| ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol | |
| CHEBI:30742 | |
| C(CO)O |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only