missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, 95%, pure

Catalog No. p-4509537
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410010010 1 kg Glass Bottle
AC410010025 2.5 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC410010010 Supplier Thermo Scientific Chemicals Supplier No. 410010010
Only null left

CAS: 107-21-1 | C2H6O2 | 62.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13.0°C
Density 1.1130g/mL
Quantity 1 kg
Flash Point 116°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Viscous Liquid
Refractive Index 1.4300 to 1.4330
Specific Gravity 1.113
Assay Percent Range 95%
Linear Formula HOCH2CH2OH
Viscosity 21 mPa.s (20°C)
Fieser 01,375; 05,296; 09,217; 15,156
Boiling Point 196.0°C to 198.0°C
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
Formula Weight 62.06
Percent Purity 95%
Grade Pure
Chemical Name or Material Ethylene glycol
Show More Show Less
Hazard Category Warning

RUO – Research Use Only