missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Glycol 1,2-Bis(2-propynyl) Ether 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 88°C |
| Quantity | 5 g |
| UN Number | 3271 |
| Formula Weight | 138.17 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Ethylene Glycol 1,2-Bis(2-propynyl) Ether |
Safety and Handling
| TSCA | No |