missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene dimethacrylate, 98%, stabilized
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -40.0°C |
| Color | Colorless to Yellow |
| Density | 1.0500g/mL |
| Boiling Point | 98.0°C to 100.0°C (5.0 mmHg) |
| Flash Point | 101°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | (H2C=C(CH3)COOCH2-)2 |
| Refractive Index | 1.4530 to 1.4550 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause an allergic skin reaction. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing. IF ON SKIN: Wash with plenty of soap and water. |
| EINECSNumber | 202-617-2 |
| RTECSNumber | OZ4400000 |
| TSCA | TSCA |
RUO – Research Use Only