missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Di(thiotosylate) 98.0+%, TCI America™

Catalog No. p-7129016
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04715G 5 g
E047125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E04715G Supplier TCI America Supplier No. E04715G
Only null left

Chemical Identifiers

CAS 2225-23-2
Molecular Formula C16H18O4S4
Molecular Weight (g/mol) 402.56
MDL Number MFCD00008550
InChI Key DUFUGAKEFZRFEQ-UHFFFAOYSA-N
PubChem CID 75212
IUPAC Name 1-methyl-4-[({2-[(4-methylbenzenesulfonyl)sulfanyl]ethyl}sulfanyl)sulfonyl]benzene
SMILES CC1=CC=C(C=C1)S(=O)(=O)SCCSS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Melting Point 74°C
Color White
Quantity 5 g
Synonym 1,2-Di(p-tosylthio)ethane, 1,2-Ethanedithiol Ditosylate, Ethylene Bis(p-toluenethiosulfonate), S,S′C-Ethylene p-Toluenethiosulfonate
Formula Weight 402.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Di(thiotosylate) [Protecting Reagent for Active Methylene]