missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Carbonate 99.0+%, TCI America™

Catalog No. E007625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E007625G 25 g
E0076500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E007625G Supplier TCI America Supplier No. E007625G

Chemical Identifiers

CAS 96-49-1
Molecular Formula C3H4O3
Molecular Weight (g/mol) 88.062
MDL Number MFCD00005382
InChI Key KMTRUDSVKNLOMY-UHFFFAOYSA-N
Synonym ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec
PubChem CID 7303
IUPAC Name 1,3-dioxolan-2-one
SMILES C1COC(=O)O1

Specifications

Melting Point 38°C
Color White
Boiling Point 244°C
Quantity 25 g
Formula Weight 88.06
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Ethylene Carbonate
EINECSNumber (5)-0523&(7)-0737
RTECSNumber FF9550000
TSCA Yes
Recommended Storage Refrigerator