missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-chloro-3,4-dimethoxybenzoate, 97%, Thermo Scientific™

Catalog No. btb09278ea
Change view
Click to view available options
Quantity:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09278EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB09278EA Supplier Thermo Scientific Chemicals Supplier No. BTB09278EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 175136-04-6
Molecular Formula C11H13ClO4
Molecular Weight (g/mol) 244.67
MDL Number MFCD00085062
InChI Key AXNMDQHZRJDVRG-UHFFFAOYSA-N
Synonym maybridge1_002113
PubChem CID 2774987
IUPAC Name ethyl 2-chloro-3,4-dimethoxybenzoate
SMILES CCOC(=O)C1=C(Cl)C(OC)=C(OC)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10g
Formula Weight 244.67
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-3, 4-dimethoxybenzoate
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.