accessibility menu, dialog, popup

UserName

H3COOOCH3OCH3Cl

CAS RN 175136-04-6

IUPAC Name: ethyl 2-chloro-3,4-dimethoxybenzoate
Synonyms: ethyl 2-chloro-3,4-dimethoxybenzoate
Molecular Weight (g/mol): 244.67
Molecular Formula: C11H13ClO4
InChi Key: AXNMDQHZRJDVRG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(Cl)C(OC)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)