missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(2-methyl-1,3-thiazol-4-yl)acetate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CD09887DE

 View more versions of this product

Catalog No. CD09887DE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

Ethyl 2-(2-methyl-1, 3-thiazol-4-yl)acetate
94.5
95%
C8H11NO2S
5g
DWSMURKWFWKPBT-UHFFFAOYSA-N
ethyl 2-(2-methyl-1,3-thiazol-4-yl)acetate
854815
95%
37128-24-8
100.0
Amber glass bottle
MFCD00052519
ethyl 2-2-methyl-1,3-thiazol-4-yl acetate, 2-methyl-thiazol-4-yl-acetic acid ethyl ester, 2-methylthiazole-4-acetic acid ethyl ester, ethyl 2-methyl-1,3-thiazol-4-yl acetate, ethyl 2-2-methylthiazol-4-yl acetate, 4-thiazoleacetic acid,2-methyl-, ethyl ester, enamine_005349, ethyl=2-methylthiazol-4-ylacetate, 2-methylthiazol-4-yl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-methyl-, ethyl ester
CCOC(=O)CC1=CSC(=N1)C
185.241
185.25

Chemical Identifiers

37128-24-8
185.241
DWSMURKWFWKPBT-UHFFFAOYSA-N
854815
CCOC(=O)CC1=CSC(=N1)C
C8H11NO2S
MFCD00052519
ethyl 2-2-methyl-1,3-thiazol-4-yl acetate, 2-methyl-thiazol-4-yl-acetic acid ethyl ester, 2-methylthiazole-4-acetic acid ethyl ester, ethyl 2-methyl-1,3-thiazol-4-yl acetate, ethyl 2-2-methylthiazol-4-yl acetate, 4-thiazoleacetic acid,2-methyl-, ethyl ester, enamine_005349, ethyl=2-methylthiazol-4-ylacetate, 2-methylthiazol-4-yl acetic acid ethyl ester, 4-thiazoleacetic acid, 2-methyl-, ethyl ester
ethyl 2-(2-methyl-1,3-thiazol-4-yl)acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS missing translation for 'documents'

missing translation for 'documents'

Promotions

Promotions