missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylcellulose, 10 mPa.s, NF, 44-51%, Spectrum™ Chemical
ET110, 9004-57-3
$575.75 - $36813.96
Chemical Identifiers
| CAS | 9004-57-3 |
|---|---|
| Molecular Formula | C23H24N6O4 |
| Molecular Weight (g/mol) | 448.48 |
| InChI Key | ARSKJXYLLONUAJ-UHFFFAOYSA-N |
| IUPAC Name | 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate |
| SMILES | CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18602554
|
Spectrum Chemical Mfg Cor
ET110125GM |
125 g | Poly Bottle |
Each for $575.75
|
|
||||
|
18602555
|
Spectrum Chemical Mfg Cor
ET110500GM |
500 g | Poly Bottle |
Each for $1,376.75
|
|
||||
|
18602556
|
Spectrum Chemical Mfg Cor
ET11025KG |
2.5 kg | Poly Pail |
Each for $4,435.54
|
|
||||
|
18602557
|
Spectrum Chemical Mfg Cor
ET11012KG |
12 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602558
|
Spectrum Chemical Mfg Cor
ET11045KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Ethylcellulose, 10 mPa.s, NF is used as a coating agent, flavoring fixative, tablet binder and filler, film-former, and as a viscosity-increasing agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 9004-57-3 | |
| 448.48 | |
| 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate |
| C23H24N6O4 | |
| ARSKJXYLLONUAJ-UHFFFAOYSA-N | |
| CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N |
Specifications
| 9004-57-3 | |
| C23H24N6O4 | |
| ARSKJXYLLONUAJ-UHFFFAOYSA-N | |
| 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate | |
| 44 to 51% | |
| 0.5% | |
| Poly Bottle |
| 100% | |
| 125 g | |
| CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N | |
| 448.48 | |
| NF | |
| 3% |