missing translation for 'onlineSavingsMsg'
Learn More

Ethylcellulose, 10 mPa.s, NF, 44-51%, Spectrum™ Chemical

Catalog No. 18602554
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602554 125 g Poly Bottle
18602555 500 g Poly Bottle
18602556 2.5 kg Poly Pail
18602557 12 kg Fiber Drum
18602558 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602554 Supplier Spectrum Chemical Mfg Cor Supplier No. ET110125GM

ET110, 9004-57-3

Spectrum™ Chemical Ethylcellulose, 10 mPa.s, NF is used as a coating agent, flavoring fixative, tablet binder and filler, film-former, and as a viscosity-increasing agent. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-57-3
Molecular Formula C23H24N6O4
Molecular Weight (g/mol) 448.48
InChI Key ARSKJXYLLONUAJ-UHFFFAOYSA-N
IUPAC Name 2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
SMILES CCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 44 to 51%
Grade NF
Ignition Residue 0.5%
Loss on Drying 3%
Packaging Poly Bottle