missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylboronic Acid (contains varying amounts of Anhydride), TCI America™
$75.08 - $75.08
Chemical Identifiers
| CAS | 4433-63-0 |
|---|---|
| Molecular Formula | C2H7BO2 |
| Molecular Weight (g/mol) | 73.89 |
| MDL Number | MFCD01074536 |
| InChI Key | PAVZHTXVORCEHP-UHFFFAOYSA-N |
| PubChem CID | 521157 |
| IUPAC Name | ethylboronic acid |
| SMILES | CCB(O)O |
Chemical Identifiers
| 4433-63-0 | |
| 73.89 | |
| PAVZHTXVORCEHP-UHFFFAOYSA-N | |
| ethylboronic acid |
| C2H7BO2 | |
| MFCD01074536 | |
| 521157 | |
| CCB(O)O |
Specifications
| 4433-63-0 | |
| C2H7BO2 | |
| 1 g | |
| PAVZHTXVORCEHP-UHFFFAOYSA-N | |
| ethylboronic acid | |
| 521157 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD01074536 | |
| boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2 | |
| CCB(O)O | |
| 73.89 | |
| 73.89 | |
| Ethylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No