Learn More
Ethylboronic acid, 97%
CAS: 4433-63-0 | C2H7BO2 | 73.89 g/mol
$89.86 - $1082.14
Chemical Identifiers
| CAS | 4433-63-0 |
|---|---|
| Molecular Formula | C2H7BO2 |
| Molecular Weight (g/mol) | 73.89 |
| MDL Number | MFCD01074536 |
| InChI Key | PAVZHTXVORCEHP-UHFFFAOYSA-N |
| Synonym | boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2 |
| PubChem CID | 521157 |
| IUPAC Name | ethylboronic acid |
| SMILES | CCB(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1995903
|
Thermo Scientific Chemicals
L1995903 |
1 g |
Each for $89.86
|
|
|||||
|
AAL1995906
|
Thermo Scientific Chemicals
L1995906 |
5 g |
Each for $273.62
|
|
|||||
|
AAL1995914
|
Thermo Scientific Chemicals
L1995914 |
25 g |
Each for $1,082.14
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4433-63-0 | |
| 73.89 | |
| PAVZHTXVORCEHP-UHFFFAOYSA-N | |
| 521157 | |
| CCB(O)O |
| C2H7BO2 | |
| MFCD01074536 | |
| boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2 | |
| ethylboronic acid |
Specifications
| 4433-63-0 | |
| Odorless | |
| CH3CH2B(OH)2 | |
| 1 g | |
| Air sensitive | |
| PAVZHTXVORCEHP-UHFFFAOYSA-N | |
| ethylboronic acid | |
| 521157 | |
| 98% |
| 84°C to 88°C | |
| C2H7BO2 | |
| MFCD01074536 | |
| 1731546 | |
| boronic acid, ethyl, ethaneboronic acid, ethyldihydroxyborane, ethyl boronic acid, boronic acid, ethyl-9ci, ethylboronicacid, pubchem7960, ethylboric acid, etb oh 2 | |
| CCB(O)O | |
| 73.89 | |
| 73.89 | |
| Ethylboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only