missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylbenzene 99.0+%, TCI America™
Supplier: TCI America E006425ML
Specifications
| Ethylbenzene | |
| Colorless | |
| C8H10 | |
| 25 mL | |
| phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene | |
| CCC1=CC=CC=C1 | |
| 106.168 | |
| CHEBI:16101 | |
| ≥99.0% (GC) |
| 100-41-4 | |
| 136°C | |
| MFCD00011647 | |
| 1175 | |
| YNQLUTRBYVCPMQ-UHFFFAOYSA-N | |
| ethylbenzene | |
| 7500 | |
| 106.17 | |
| Liquid |
Chemical Identifiers
| 100-41-4 | |
| 106.168 | |
| YNQLUTRBYVCPMQ-UHFFFAOYSA-N | |
| 7500 | |
| ethylbenzene |
| C8H10 | |
| MFCD00011647 | |
| phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene | |
| CHEBI:16101 | |
| CCC1=CC=CC=C1 |
Safety and Handling
EINECSNumber : (3)-0028&(3)-0060
RTECSNumber : DA0700000
TSCA : Yes