missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene 99.0+%, TCI America™

Catalog No. p-7128818
Change view
Click to view available options
Quantity:
25 mL
500 mL
Catalog No. Quantity
E006425ML 25 mL
E0064500ML 500 mL
2 options

Catalog No. E006425ML

Supplier: TCI America E006425ML

Only null left

Chemical Identifiers

CAS 100-41-4
Molecular Formula C8H10
Molecular Weight (g/mol) 106.168
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 136°C
Quantity 25 mL
UN Number 1175
Formula Weight 106.17
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethylbenzene
EINECSNumber (3)-0028&(3)-0060
RTECSNumber DA0700000
TSCA Yes