missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023058
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023058 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023058 Supplier Merck Emd Millipore Supplier No. RTC00010020ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 100-41-4
Vapor Pressure 19 mmHg (37.7°C)
Vapor Density 3.7 (vs air)
Density 0.867 g/mL (at 25°C (literature))
Flash Point 15°C (Not Applicable)
Molecular Formula C8H10
Linear Formula C6H5C2H5
MDL Number MFCD00011647
Melting Point -95°C (literature)
Boiling Point 136°C (literature)
Synonym Ethylbenzene; NSC 406903; Phenylethane
Quantity 20 mL
Formula Weight 106.17
Grade Certified Reference Material
Show More Show Less