missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Violet, TCI America™

Catalog No. E020025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E020025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E020025G Supplier TCI America Supplier No. E020025G

Chemical Identifiers

CAS 2390-59-2
Molecular Formula C31H42ClN3
Molecular Weight (g/mol) 492.15
MDL Number MFCD00011765
InChI Key JVICFMRAVNKDOE-UHFFFAOYSA-M
Synonym ethyl violet, basic violet 4, c.i. basic violet 4, ethyl violet ax, ethyl violet gga, ethyl crystal violet, lowacryl violet 4, shikiso acid brilliant blue 6b, ethyl violet biological stain, unii-94uz9rd7tj
PubChem CID 16955
IUPAC Name tris[4-(diethylamino)phenyl]methylium chloride
SMILES [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC

Specifications

Quantity 25 g
Formula Weight 560.29
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Violet
RTECSNumber KH2682000
TSCA Yes