missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Vinyl Ether (stabilized with KOH) 98.0+%, TCI America™
Supplier: TCI America E0193100ML
Specifications
| Ethyl Vinyl Ether (stabilized with KOH) | |
| -115°C | |
| 36°C | |
| MFCD00009248 | |
| 1302 | |
| FJKIXWOMBXYWOQ-UHFFFAOYSA-N | |
| ethoxyethene | |
| 8023 | |
| ≥98.0% (GC) |
| 109-92-2 | |
| Colorless | |
| C4H8O | |
| 100 mL | |
| ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl | |
| CCOC=C | |
| 72.11 | |
| 72.11 | |
| Liquid |
Chemical Identifiers
| 109-92-2 | |
| 72.11 | |
| FJKIXWOMBXYWOQ-UHFFFAOYSA-N | |
| 8023 | |
| CCOC=C |
| C4H8O | |
| MFCD00009248 | |
| ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl | |
| ethoxyethene |
Safety and Handling
EINECSNumber : (2)-0372
RTECSNumber : KO0710000
TSCA : Yes
Recommended Storage : Refrigerator