accessibility menu, dialog, popup

UserName

CH3OH2C

CAS RN 109-92-2

IUPAC Name: ethoxyethene
Synonyms: ethyl vinyl ether
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: FJKIXWOMBXYWOQ-UHFFFAOYSA-N
SMILES: CCOC=C
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)