missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Vinyl Ether (stabilized with KOH) 98.0+%, TCI America™

Catalog No. E0193500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0193500ML 500 mL
E019325ML 25 mL
E0193100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0193500ML Supplier TCI America Supplier No. E0193500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-92-2
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00009248
InChI Key FJKIXWOMBXYWOQ-UHFFFAOYSA-N
Synonym ethyl vinyl ether, ethoxyethene, ethoxyethylene, ethene, ethoxy, 1-ethoxyethene, 1-ethoxyethylene, vinyl ethyl ether, vinamar, ether, ethyl vinyl, ether, vinyl ethyl
PubChem CID 8023
IUPAC Name ethoxyethene
SMILES CCOC=C

Specifications

Melting Point -115°C
Color Colorless
Boiling Point 36°C
Quantity 500 mL
UN Number 1302
Formula Weight 72.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Vinyl Ether (stabilized with KOH)
EINECSNumber (2)-0372
RTECSNumber KO0710000
TSCA Yes
Recommended Storage Refrigerator