missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Valerate 98.0+%, TCI America™
$41.02 - $123.03
Chemical Identifiers
| CAS | 539-82-2 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.187 |
| MDL Number | MFCD00009479 |
| InChI Key | ICMAFTSLXCXHRK-UHFFFAOYSA-N |
| Synonym | ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462 |
| PubChem CID | 10882 |
| IUPAC Name | ethyl pentanoate |
| SMILES | CCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
V000425ML
|
TCI America
V000425ML |
25 mL |
Each for $41.02
|
|
|||||
|
V0004500ML
|
TCI America
V0004500ML |
500 mL |
Each for $123.03
|
|
|||||
Chemical Identifiers
| 539-82-2 | |
| 130.187 | |
| ICMAFTSLXCXHRK-UHFFFAOYSA-N | |
| 10882 | |
| CCCCC(=O)OCC |
| C7H14O2 | |
| MFCD00009479 | |
| ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462 | |
| ethyl pentanoate |
Specifications
| 539-82-2 | |
| 145°C | |
| MFCD00009479 | |
| 3272 | |
| ICMAFTSLXCXHRK-UHFFFAOYSA-N | |
| ethyl pentanoate | |
| 10882 | |
| ≥98.0% (GC) | |
| Ethyl Valerate |
| Colorless | |
| C7H14O2 | |
| 25 mL | |
| ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462 | |
| CCCCC(=O)OCC | |
| 130.187 | |
| 130.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0776
TSCA : Yes