missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Valerate 98.0+%, TCI America™

Catalog No. V000425ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V000425ML 25 mL
V0004500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. V000425ML Supplier TCI America Supplier No. V000425ML

Chemical Identifiers

CAS 539-82-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009479
InChI Key ICMAFTSLXCXHRK-UHFFFAOYSA-N
Synonym ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
PubChem CID 10882
IUPAC Name ethyl pentanoate
SMILES CCCCC(=O)OCC

Specifications

Color Colorless
Boiling Point 145°C
Quantity 25 mL
UN Number 3272
Formula Weight 130.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Valerate
EINECSNumber (2)-0776
TSCA Yes