missing translation for 'onlineSavingsMsg'
Learn More

Ethyl valerate, 99%

Catalog No. AC410470050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC410470050 5 g Glass bottle
AC410471000 100 g Glass bottle
AC410475000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC410470050 Supplier Thermo Scientific Chemicals Supplier No. 410470050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 539-82-2 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 539-82-2
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009479
InChI Key ICMAFTSLXCXHRK-UHFFFAOYSA-N
Synonym ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462
PubChem CID 10882
IUPAC Name ethyl pentanoate
SMILES CCCCC(=O)OCC

Specifications

Color Colorless
Density 0.8700g/mL
Boiling Point 145°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.3990 to 1.401
Linear Formula CH3(CH2)3COOC2H5
Quantity 5 g
Specific Gravity 0.87
Merck Index 15, 10090
Formula Weight 130.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethyl valerate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-726-1

RUO – Research Use Only