Learn More
Ethyl valerate, 98%
CAS: 539-82-2 | C7H14O2 | 130.187 g/mol
$123.93 - $522.53
Chemical Identifiers
| CAS | 539-82-2 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.187 |
| MDL Number | MFCD00009479 |
| InChI Key | ICMAFTSLXCXHRK-UHFFFAOYSA-N |
| Synonym | ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462 |
| PubChem CID | 10882 |
| IUPAC Name | ethyl pentanoate |
| SMILES | CCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA14854AP
|
Thermo Scientific Chemicals
A14854AP |
500 mL |
Each for $123.93
|
|
|||||
|
AAA148540F
|
Thermo Scientific Chemicals
A148540F |
2500 mL |
Each for $522.53
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 539-82-2 | |
| 130.187 | |
| ICMAFTSLXCXHRK-UHFFFAOYSA-N | |
| 10882 | |
| CCCCC(=O)OCC |
| C7H14O2 | |
| MFCD00009479 | |
| ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462 | |
| ethyl pentanoate |
Specifications
| 539-82-2 | |
| 0.874 | |
| 38°C (100°F) | |
| 1.4 | |
| 500 mL | |
| 1744680 | |
| ethyl valerate, ethyl n-valerate, pentanoic acid, ethyl ester, valeric acid, ethyl ester, ethylvalerate, ethyl valerianate, pentanoic acid ethyl ester, n-valeric acid ethyl ester, valeric acid, ethyl ester 8ci, fema no. 2462 | |
| CCCCC(=O)OCC | |
| 130.187 | |
| 130.19 | |
| Ethyl valerate |
| -92°C to -90°C | |
| 142°C to 145°C | |
| C7H14O2 | |
| MFCD00009479 | |
| UN3272 | |
| 14,9904 | |
| ICMAFTSLXCXHRK-UHFFFAOYSA-N | |
| ethyl pentanoate | |
| 10882 | |
| 98% |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 208-726-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only