missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (trimethylsilyl)acetate, 98%

CAS: 4071-88-9 | C7H16O2Si | 160.288 g/mol

Supplier:  Thermo Scientific Chemicals A1770718

 View more versions of this product

Catalog No. AAA1770718


Only null left
Explore available promotions

Description

Description

Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Ethyl (trimethylsilyl)acetate
0.876
35°C (95°F)
1.415
50 g
1755902
ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie
QQFBQBDINHJDMN-UHFFFAOYSA-N
ethyl 2-trimethylsilylacetate
77687
≥98%
4071-88-9
156°C to 158°C
C7H16O2Si
MFCD00009172
UN3272
Moisture sensitive
Decomposes in water.
CCOC(=O)C[Si](C)(C)C
160.288
160.29

Chemical Identifiers

4071-88-9
160.288
QQFBQBDINHJDMN-UHFFFAOYSA-N
77687
CCOC(=O)C[Si](C)(C)C
C7H16O2Si
MFCD00009172
ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie
ethyl 2-trimethylsilylacetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 223-783-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only