missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (trimethylsilyl)acetate, 98%

Catalog No. p-7023800
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1770709 10 g
AAA1770718 50 g
AAA1770730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1770709 Supplier Thermo Scientific Chemicals Supplier No. A1770709
Only null left

CAS: 4071-88-9 | C7H16O2Si | 160.288 g/mol

Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl trimethylsilylacetate is used in the synthesis of ethyl-2,2-dibromo-2-(trimethylsilyl)acetate. It was also used to silylate the enolizable aldehydes and ketones.

Solubility
Decomposes in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 4071-88-9
Molecular Formula C7H16O2Si
Molecular Weight (g/mol) 160.288
MDL Number MFCD00009172
InChI Key QQFBQBDINHJDMN-UHFFFAOYSA-N
Synonym ethyl trimethylsilyl acetate, ethyl trimethylsilylacetate, ethyl 2-trimethylsilyl acetate, etsa, trimethylsilyl acetic acid ethyl ester, acetic acid, trimethylsilyl-, ethyl ester, pubchem16058, acmc-1ahie
PubChem CID 77687
IUPAC Name ethyl 2-trimethylsilylacetate
SMILES CCOC(=O)C[Si](C)(C)C

Specifications

Density 0.876
Boiling Point 156°C to 158°C
Flash Point 35°C (95°F)
Refractive Index 1.415
Quantity 10 g
UN Number UN3272
Beilstein 1755902
Sensitivity Moisture sensitive
Solubility Information Decomposes in water.
Formula Weight 160.29
Percent Purity ≥98%
Chemical Name or Material Ethyl (trimethylsilyl)acetate
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber 223-783-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only