missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trimethylacetopyruvate, 98%

Catalog No. AAA1516406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1516406 5 g
AAA1516403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1516406 Supplier Thermo Scientific Chemicals Supplier No. A1516406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13395-36-3 | C10H16O4 | 200.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13395-36-3
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00052319
InChI Key NIMKIMUBJFWPTD-UHFFFAOYSA-N
Synonym ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
PubChem CID 83403
IUPAC Name ethyl 5,5-dimethyl-2,4-dioxohexanoate
SMILES CCOC(=O)C(=O)CC(=O)C(C)(C)C

Specifications

Boiling Point 68°C to 70°C (0.5 mmHg)
Refractive Index 1.47
Quantity 5 g
Beilstein 510374
Formula Weight 200.24
Percent Purity 98%
Chemical Name or Material Ethyl trimethylacetopyruvate
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-478-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only