missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7136649
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
T11915G 5 g
T119125G 25 g
2 options

Catalog No. T11915G

Supplier: TCI America T11915G

Only null left

Chemical Identifiers

CAS 425-75-2
Molecular Formula C3H5F3O3S
Molecular Weight (g/mol) 178.125
MDL Number MFCD00000410
InChI Key UVECLJDRPFNRRQ-UHFFFAOYSA-N
Synonym ethyl triflate, ethyl trifluoromethanesulphonate, trifluoromethanesulfonic acid ethyl ester, methanesulfonic acid, trifluoro-, ethyl ester, methanesulfonic acid, 1,1,1-trifluoro-, ethyl ester, ethyl trifluoromethyl sulfonate, ethyl triflate∼trifluoromethanesulphonic acid ethyl ester, ethyl trifiate, etotf, acmc-209jqf
PubChem CID 67924
IUPAC Name ethyl trifluoromethanesulfonate
SMILES CCOS(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 115°C
Quantity 5 g
UN Number 2924
Formula Weight 178.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Trifluoromethanesulfonate