accessibility menu, dialog, popup

UserName

CH3OSOOFFF

CAS RN 425-75-2

IUPAC Name: ethyl trifluoromethanesulfonate
Synonyms: ethyl trifluoromethanesulfonate
Molecular Weight (g/mol): 178.13
Molecular Formula: C3H5F3O3S
InChi Key: UVECLJDRPFNRRQ-UHFFFAOYSA-N
SMILES: CCOS(=O)(=O)C(F)(F)F
Grade 
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results