missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Tridecanoate 97.0+%, TCI America™
$162.17 - $492.11
Chemical Identifiers
| CAS | 28267-29-0 |
|---|---|
| Molecular Formula | C15H30O2 |
| Molecular Weight (g/mol) | 242.403 |
| MDL Number | MFCD00008978 |
| InChI Key | QJYYMNOTJXIOBP-UHFFFAOYSA-N |
| Synonym | tridecanoic acid ethyl ester, tridecanoic acid, ethyl ester, ethyltridecanoate, ethyl=tridecanoate, tridecanoic acid, ethylester, acmc-1cr51, ethyl tridecanoate, n-tridecanoic acid ethyl ester |
| PubChem CID | 119908 |
| IUPAC Name | ethyl tridecanoate |
| SMILES | CCCCCCCCCCCCC(=O)OCC |
Chemical Identifiers
| 28267-29-0 | |
| 242.403 | |
| QJYYMNOTJXIOBP-UHFFFAOYSA-N | |
| 119908 | |
| CCCCCCCCCCCCC(=O)OCC |
| C15H30O2 | |
| MFCD00008978 | |
| tridecanoic acid ethyl ester, tridecanoic acid, ethyl ester, ethyltridecanoate, ethyl=tridecanoate, tridecanoic acid, ethylester, acmc-1cr51, ethyl tridecanoate, n-tridecanoic acid ethyl ester | |
| ethyl tridecanoate |
Specifications
| 28267-29-0 | |
| 198°C | |
| MFCD00008978 | |
| tridecanoic acid ethyl ester, tridecanoic acid, ethyl ester, ethyltridecanoate, ethyl=tridecanoate, tridecanoic acid, ethylester, acmc-1cr51, ethyl tridecanoate, n-tridecanoic acid ethyl ester | |
| CCCCCCCCCCCCC(=O)OCC | |
| 242.403 | |
| 242.40 | |
| Liquid |
| Colorless | |
| C15H30O2 | |
| 5 mL | |
| QJYYMNOTJXIOBP-UHFFFAOYSA-N | |
| ethyl tridecanoate | |
| 119908 | |
| ≥97.0% (GC) | |
| Ethyl Tridecanoate |
Safety and Handling
EINECSNumber : (2)-0798
TSCA : Yes