accessibility menu, dialog, popup

UserName

CAS RN 28267-29-0

IUPAC Name: ethyl tridecanoate
Synonyms: ethyl tridecanoate
Molecular Weight (g/mol): 242.4
Molecular Formula: C15H30O2
InChi Key: QJYYMNOTJXIOBP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCC(=O)OCC
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)