missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Tridecanoate 97.0+%, TCI America™

Catalog No. T09595ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09595ML 5 mL
T095925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09595ML Supplier TCI America Supplier No. T09595ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28267-29-0
Molecular Formula C15H30O2
Molecular Weight (g/mol) 242.403
MDL Number MFCD00008978
InChI Key QJYYMNOTJXIOBP-UHFFFAOYSA-N
Synonym tridecanoic acid ethyl ester, tridecanoic acid, ethyl ester, ethyltridecanoate, ethyl=tridecanoate, tridecanoic acid, ethylester, acmc-1cr51, ethyl tridecanoate, n-tridecanoic acid ethyl ester
PubChem CID 119908
IUPAC Name ethyl tridecanoate
SMILES CCCCCCCCCCCCC(=O)OCC

Specifications

Color Colorless
Boiling Point 198°C
Quantity 5 mL
Formula Weight 242.40
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Tridecanoate
EINECSNumber (2)-0798
TSCA Yes