missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-4-Decenoate 98.0+%, TCI America™

Catalog No. D193110ML
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D193110ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D193110ML Supplier TCI America Supplier No. D193110ML

Chemical Identifiers

CAS 76649-16-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00015574
InChI Key AWNIQMQADACLCJ-CMDGGOBGSA-N
Synonym ethyl trans-4-decenoate, ethyl e-4-decenoate, ethyl 4-decenoate, 4-decenoic acid, ethyl ester, e, ethyl 4e-decenoate, fema no. 3642, ethyl 4e-dec-4-enoate, ethyl trans-dec-4-enoate, ethyl e-dec-4-enoate, ethyl 4e-4-decenoate
PubChem CID 5362583
IUPAC Name ethyl (4E)-dec-4-enoate
SMILES CCCCC\C=C\CCC(=O)OCC

Specifications

Color Colorless
Boiling Point 70°C
Quantity 10 mL
Formula Weight 198.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl trans-4-Decenoate