accessibility menu, dialog, popup

UserName

H3COOCH3(E)

CAS RN 76649-16-6

IUPAC Name: ethyl (4E)-dec-4-enoate
Synonyms: ethyl (4E)-dec-4-enoate
Molecular Weight (g/mol): 198.31
Molecular Formula: C12H22O2
InChi Key: AWNIQMQADACLCJ-CMDGGOBGSA-N
SMILES: CCCCC\C=C\CCC(=O)OCC
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results