missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-4-decenoate, 97%

Catalog No. AAA1936409
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1936409 10 g
AAA1936418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1936409 Supplier Thermo Scientific Chemicals Supplier No. A1936409
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 76649-16-6 | C12H22O2 | 198.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 76649-16-6
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.31
MDL Number MFCD00015574
InChI Key AWNIQMQADACLCJ-CMDGGOBGSA-N
Synonym ethyl trans-4-decenoate, ethyl e-4-decenoate, ethyl 4-decenoate, 4-decenoic acid, ethyl ester, e, ethyl 4e-decenoate, fema no. 3642, ethyl 4e-dec-4-enoate, ethyl trans-dec-4-enoate, ethyl e-dec-4-enoate, ethyl 4e-4-decenoate
PubChem CID 5362583
IUPAC Name ethyl (E)-dec-4-enoate
SMILES CCCCC\C=C\CCC(=O)OCC

Specifications

Refractive Index 1.437
Quantity 10 g
Formula Weight 198.31
Percent Purity 97%
Chemical Name or Material Ethyl trans-4-decenoate
EINECSNumber 278-509-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only