missing translation for 'onlineSavingsMsg'
Learn More

Ethyl trans-4-bromocinnamate, 98%

Catalog No. AAH2754414
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2754414 25 g
AAH2754406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2754414 Supplier Thermo Scientific Chemicals Supplier No. H2754414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24393-53-1 | C11H11BrO2 | 255.111 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 24393-53-1
Molecular Formula C11H11BrO2
Molecular Weight (g/mol) 255.111
MDL Number MFCD00012226
InChI Key YOOKYIPLSLPRTC-VMPITWQZSA-N
Synonym ethyl trans-4-bromocinnamate, e-ethyl 3-4-bromophenyl acrylate, ethyl 3-4-bromophenyl acrylate, 2-propenoic acid, 3-4-bromophenyl-, ethyl ester, ethyl 2e-3-4-bromophenyl prop-2-enoate, 2-propenoic acid, 3-4-bromophenyl-, ethyl ester, 2e, ethyl p-bromocinnamate, ethyl e-4-bromocinnamate, ethyl-trans-4-bromocinnamate, 4-bromocinnamic acid ethyl ester
PubChem CID 688115
IUPAC Name ethyl (E)-3-(4-bromophenyl)prop-2-enoate
SMILES CCOC(=O)C=CC1=CC=C(C=C1)Br

Specifications

Density 1.359
Boiling Point 180°C (18 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.597
Quantity 25 g
Formula Weight 255.11
Percent Purity 98%
Chemical Name or Material Ethyl trans-4-bromocinnamate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only