CAS RN 24393-53-1
IUPAC Name:
ethyl (2E)-3-(4-bromophenyl)prop-2-enoate
Synonyms:
ethyl (2E)-3-(4-bromophenyl)prop-2-enoate
Molecular Weight (g/mol):
255.11
Molecular Formula:
C11H11BrO2
InChi Key:
YOOKYIPLSLPRTC-VMPITWQZSA-N
SMILES:
CCOC(=O)\C=C\C1=CC=C(Br)C=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl trans-4-bromocinnamate, 98%
Ethyl trans-4-bromocinnamate, 98%
CAS: 24393-53-1 Molecular Formula: C11H11BrO2 Molecular Weight (g/mol): 255.111 MDL Number: MFCD00012226 InChI Key: YOOKYIPLSLPRTC-VMPITWQZSA-N Synonym: ethyl trans-4-bromocinnamate,e-ethyl 3-4-bromophenyl acrylate,ethyl 3-4-bromophenyl acrylate,2-propenoic acid, 3-4-bromophenyl-, ethyl ester,ethyl 2e-3-4-bromophenyl prop-2-enoate,2-propenoic acid, 3-4-bromophenyl-, ethyl ester, 2e,ethyl p-bromocinnamate,ethyl e-4-bromocinnamate,ethyl-trans-4-bromocinnamate,4-bromocinnamic acid ethyl ester PubChem CID: 688115 IUPAC Name: ethyl (E)-3-(4-bromophenyl)prop-2-enoate SMILES: CCOC(=O)C=CC1=CC=C(C=C1)Br