missing translation for 'onlineSavingsMsg'
Learn More

Ethyl thiophene-2-glyoxylate, 97%

Catalog No. p-7027727
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2471209 10 g
AAB2471218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2471209 Supplier Thermo Scientific Chemicals Supplier No. B2471209

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4075-58-5 | C8H8O3S | 184.209 g/mol

Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl thiophene-2-glyoxylate is used for drug discovery using chemic informatics-based design and it is used as a catalyst, synthesis of raw material.

Solubility
Difficult to mix.

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 4075-58-5
Molecular Formula C8H8O3S
Molecular Weight (g/mol) 184.209
MDL Number MFCD00015538
InChI Key GHOVLEQTRNXASK-UHFFFAOYSA-N
Synonym ethyl thiophene-2-glyoxylate, ethyl 2-oxo-2-thiophen-2-yl acetate, ethyl 2-oxo-2-2-thienyl acetate, ethyl alpha-oxothiophen-2-acetate, ethyl 2-thienylglyoxylate, thiophen-2-coco-o-ethyl, 2-thiophene glyoxylic acid ethyl ester, oxo-thiophen-2-yl-acetic acid ethyl ester, 2-thiophene glyoxalicacid ethyl ester, ethyl oxo 2-thienyl acetate
PubChem CID 77693
IUPAC Name ethyl 2-oxo-2-thiophen-2-ylacetate
SMILES CCOC(=O)C(=O)C1=CC=CS1

Specifications

Density 1.25
Boiling Point 102°C to 110°C (0.8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5505
Quantity 10 g
Beilstein 5651
Solubility Information Difficult to mix.
Formula Weight 184.21
Percent Purity 97%
Chemical Name or Material Ethyl thiophene-2-glyoxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 223-793-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only