accessibility menu, dialog, popup

UserName

CAS RN 4075-58-5

IUPAC Name: ethyl 2-oxo-2-(thiophen-2-yl)acetate
Synonyms: thiophen-2-coco-O-ethyl
Molecular Weight (g/mol): 184.21
Molecular Formula: C8H8O3S
InChi Key: GHOVLEQTRNXASK-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=CC=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results