missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Thiooxamate 98.0+%, TCI America™

$54.06 - $150.98

Chemical Identifiers

CAS 16982-21-1
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00074903
InChI Key YMBMCMOZIGSBOA-UHFFFAOYSA-N
Synonym ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
PubChem CID 2733398
IUPAC Name ethyl 2-amino-2-sulfanylideneacetate
SMILES CCOC(=O)C(=S)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
E07351G
View Documents
TCI America
E07351G
1 g
Each for $54.06
Only null left
 
E07355G
View Documents
TCI America
E07355G
5 g
Each for $150.98
Only null left
 
Description

Description

Specifications

Chemical Identifiers

16982-21-1
133.165
YMBMCMOZIGSBOA-UHFFFAOYSA-N
2733398
CCOC(=O)C(=S)N
C4H7NO2S
MFCD00074903
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
ethyl 2-amino-2-sulfanylideneacetate

Specifications

16982-21-1
C4H7NO2S
1 g
YMBMCMOZIGSBOA-UHFFFAOYSA-N
ethyl 2-amino-2-sulfanylideneacetate
2733398
≥98.0% (GC,N)
Ethyl Thiooxamate
64°C
MFCD00074903
ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
CCOC(=O)C(=S)N
133.165
133.17
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'