missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Thiooxamate 98.0+%, TCI America™

Catalog No. E07351G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07351G 1 g
E07355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07351G Supplier TCI America Supplier No. E07351G
Only null left

Chemical Identifiers

CAS 16982-21-1
Molecular Formula C4H7NO2S
Molecular Weight (g/mol) 133.165
MDL Number MFCD00074903
InChI Key YMBMCMOZIGSBOA-UHFFFAOYSA-N
Synonym ethyl 2-amino-2-thioxoacetate, ethyl carbamothioylformate, ethyl thioxamate, ethyl thiooxamidate, ethyl amino thioxo acetate, ethyl thiooxamat, ethyl aminothioxoacetate, ethyl 2-thiooxamate, acetic acid, aminothioxo-, ethyl ester, thiooxamic acid ethyl ester
PubChem CID 2733398
IUPAC Name ethyl 2-amino-2-sulfanylideneacetate
SMILES CCOC(=O)C(=S)N

Specifications

Melting Point 64°C
Quantity 1 g
Formula Weight 133.17
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material Ethyl Thiooxamate
TSCA No
Recommended Storage Refrigerator