missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Tetrahydrofuran-2-acetate 98.0+%, TCI America™

Catalog No. p-7136518
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T09575ML 5 mL
T095725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T09575ML Supplier TCI America Supplier No. T09575ML

Chemical Identifiers

CAS 2434-02-8
Molecular Formula C8H14O3
Molecular Weight (g/mol) 158.20
MDL Number MFCD00059735
InChI Key SNSYYBGUDOYAMB-UHFFFAOYNA-N
Synonym Tetrahydrofuran-2-acetic Acid Ethyl Ester
PubChem CID 551345
IUPAC Name ethyl 2-(oxolan-2-yl)acetate
SMILES CCOC(=O)CC1CCCO1

Specifications

Color Colorless
Quantity 5 mL
Physical Form Clear Liquid at 20°C
Boiling Point 98°C
Formula Weight 158.20
Percent Purity ≥98.0% (GC)
Chemical Name or Material Ethyl Tetrahydrofuran-2-acetate