accessibility menu, dialog, popup

UserName

CAS RN 2434-02-8

H3COOO

CAS RN 2434-02-8

IUPAC Name: ethyl 2-(oxolan-2-yl)acetate
Synonyms: ethyl 2-(oxolan-2-yl)acetate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: SNSYYBGUDOYAMB-UHFFFAOYNA-N
SMILES: CCOC(=O)CC1CCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results