missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Sulfide 98.0+%, TCI America™

Catalog No. p-7128878
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mL
25 mL
Catalog No. Quantity
E017625ML 25 mL
E0176100ML 100 mL
2 options

Catalog No. E017625ML

Supplier: TCI America E017625ML

Only null left

Chemical Identifiers

CAS 352-93-2
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00009270
InChI Key LJSQFQKUNVCTIA-UHFFFAOYSA-N
Synonym diethyl sulfide, ethyl sulfide, ethyl thioether, 3-thiapentane, diethylthioether, thioethyl ether, ethylthioethane, 1,1'-thiobisethane, diethyl sulphide, ethane, 1,1'-thiobis
PubChem CID 9609
ChEBI CHEBI:27710
IUPAC Name (ethylsulfanyl)ethane
SMILES CCSCC

Specifications

Color Colorless
Boiling Point 92°C
Quantity 25 mL
UN Number 2375
Formula Weight 90.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Sulfide
RTECSNumber LC7200000
TSCA Yes