missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Stearate 96.0+%, TCI America™
$72.70 - $152.86
Chemical Identifiers
| CAS | 111-61-5 |
|---|---|
| Molecular Formula | C20H40O2 |
| Molecular Weight (g/mol) | 312.538 |
| MDL Number | MFCD00009006 |
| InChI Key | MVLVMROFTAUDAG-UHFFFAOYSA-N |
| Synonym | ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate |
| PubChem CID | 8122 |
| ChEBI | CHEBI:84936 |
| IUPAC Name | ethyl octadecanoate |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)OCC |
Chemical Identifiers
| 111-61-5 | |
| 312.538 | |
| MVLVMROFTAUDAG-UHFFFAOYSA-N | |
| 8122 | |
| ethyl octadecanoate |
| C20H40O2 | |
| MFCD00009006 | |
| ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate | |
| CHEBI:84936 | |
| CCCCCCCCCCCCCCCCCC(=O)OCC |
Specifications
| 111-61-5 | |
| 199°C | |
| MFCD00009006 | |
| ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate | |
| CCCCCCCCCCCCCCCCCC(=O)OCC | |
| 312.538 | |
| CHEBI:84936 | |
| ≥96.0% (GC) | |
| Ethyl Stearate |
| White-Yellow | |
| C20H40O2 | |
| 25 g | |
| MVLVMROFTAUDAG-UHFFFAOYSA-N | |
| ethyl octadecanoate | |
| 8122 | |
| 312.54 | |
| Crystalline Lumps |
Safety and Handling
EINECSNumber : (2)-0798
RTECSNumber : WI3600000
TSCA : Yes