missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl stearate, 97%
CAS: 111-61-5 | C20H40O2 | 312.52 g/mol
$60.69 - $194.50
Chemical Identifiers
| CAS | 111-61-5 |
|---|---|
| Molecular Formula | C20H40O2 |
| Molecular Weight (g/mol) | 312.52 |
| MDL Number | MFCD00009006 |
| InChI Key | MVLVMROFTAUDAG-UHFFFAOYSA-N |
| Synonym | ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate |
| PubChem CID | 8122 |
| ChEBI | CHEBI:84936 |
| IUPAC Name | ethyl octadecanoate |
| SMILES | CCCCCCCCCCCCCCCCCC(=O)OCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC207480050
|
Thermo Scientific Chemicals
207480050 |
5 g | Glass bottle |
Each for $60.69
|
|
||||
|
AC207480250
|
Thermo Scientific Chemicals
207480250 |
25 g | Glass bottle |
Each for $194.50
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 111-61-5 | |
| 312.52 | |
| MVLVMROFTAUDAG-UHFFFAOYSA-N | |
| 8122 | |
| ethyl octadecanoate |
| C20H40O2 | |
| MFCD00009006 | |
| ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate | |
| CHEBI:84936 | |
| CCCCCCCCCCCCCCCCCC(=O)OCC |
Specifications
| 111-61-5 | |
| White | |
| >112°C | |
| 96% min. (GC) | |
| C20H40O2 | |
| MFCD00009006 | |
| 02, 379 | |
| ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate | |
| MVLVMROFTAUDAG-UHFFFAOYSA-N | |
| ethyl octadecanoate | |
| 8122 | |
| 312.52 | |
| Crystalline Mass |
| 30°C to 39°C | |
| 213°C to 215°C (15.0 mmHg) | |
| Authentic | |
| Glass bottle | |
| CH3(CH2)16CO2C2H5 | |
| 5 g | |
| 15, 8930 | |
| Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,acetone | |
| CCCCCCCCCCCCCCCCCC(=O)OCC | |
| 312.52 | |
| CHEBI:84936 | |
| 97% | |
| Ethyl stearate, 97% |
Safety and Handling
EINECSNumber : 203-887-4
RTECSNumber : WI3600000
TSCA : TSCA
RUO – Research Use Only