missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Salicylate 99.0+%, TCI America™

Catalog No. p-7135762
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
S001125ML 25 mL
S0011500ML 500 mL
2 options

Catalog No. S001125ML

Supplier: TCI America S001125ML

Only null left

Chemical Identifiers

CAS 118-61-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002215
InChI Key GYCKQBWUSACYIF-UHFFFAOYSA-N
Synonym ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
PubChem CID 8365
IUPAC Name ethyl 2-hydroxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1O

Specifications

Melting Point 1°C
Color Colorless
Boiling Point 231°C
Quantity 25 mL
Formula Weight 166.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Salicylate
EINECSNumber (3)-1585
RTECSNumber VO3000000
TSCA Yes