missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Salicylate 99.0+%, TCI America™

Catalog No. S0011500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0011500ML 500 mL
S001125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S0011500ML Supplier TCI America Supplier No. S0011500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 118-61-6
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002215
InChI Key GYCKQBWUSACYIF-UHFFFAOYSA-N
Synonym ethyl salicylate, sal ethyl, mesotol, salotan, salicylic ether, salicylic acid, ethyl ester, benzoic acid, 2-hydroxy-, ethyl ester, ethyl o-hydroxybenzoate, o-ethoxycarbonyl phenol, 2-hydroxybenzoic acid ethyl ester
PubChem CID 8365
IUPAC Name ethyl 2-hydroxybenzoate
SMILES CCOC(=O)C1=CC=CC=C1O

Specifications

Melting Point 1°C
Color Colorless
Boiling Point 231°C
Quantity 500 mL
Formula Weight 166.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethyl Salicylate
EINECSNumber (3)-1585
RTECSNumber VO3000000
TSCA Yes