missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (S)-4-Chloro-3-hydroxybutyrate 95.0+%, TCI America™
$87.60 - $158.44
Chemical Identifiers
| CAS | 86728-85-0 |
|---|---|
| Molecular Formula | C6H11ClO3 |
| Molecular Weight (g/mol) | 166.60 |
| MDL Number | MFCD00211241 |
| InChI Key | ZAJNMXDBJKCCAT-YFKPBYRVSA-N |
| Synonym | ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate |
| PubChem CID | 7019277 |
| IUPAC Name | ethyl (3S)-4-chloro-3-hydroxybutanoate |
| SMILES | CCOC(=O)C[C@H](O)CCl |
Chemical Identifiers
| 86728-85-0 | |
| 166.60 | |
| ZAJNMXDBJKCCAT-YFKPBYRVSA-N | |
| 7019277 | |
| CCOC(=O)C[C@H](O)CCl |
| C6H11ClO3 | |
| MFCD00211241 | |
| ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate | |
| ethyl (3S)-4-chloro-3-hydroxybutanoate |
Specifications
| 86728-85-0 | |
| 96°C | |
| MFCD00211241 | |
| ethyl s-4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutyrate, ethyl s---4-chloro-3-hydroxybutanoate, ethyl 3s-4-chloro-3-hydroxybutanoate, ethyl s-4-chloro-3-hydroxybutanoate, s-4-chloro-3-hydroxybutyrate, s-4-chloro-3-hydroxybutyric acid ethyl ester, s-4-chloro-3-hydroxy-n-butyric acid ethyl ester, s-ethyl 4-chloro-3-hydroxybutanoate | |
| CCOC(=O)C[C@H](O)CCl | |
| 166.60 | |
| 166.60 | |
| Liquid |
| Yellow | |
| C6H11ClO3 | |
| 5 g | |
| ZAJNMXDBJKCCAT-YFKPBYRVSA-N | |
| ethyl (3S)-4-chloro-3-hydroxybutanoate | |
| 7019277 | |
| ≥95.0% (GC) | |
| Ethyl (S)-4-Chloro-3-hydroxybutyrate |
Safety and Handling
EINECSNumber : (2)-3782&(2)-3796&(2)-3882
TSCA : No
Recommended Storage : Refrigerator